Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Undecylthiophene 98.0+%, TCI America™
SDP

Catalog No. U00561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00561G 1 g
U00565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00561G Supplier TCI America Supplier No. U00561G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129607-86-9
Molecular Formula C15H26S
Molecular Weight (g/mol) 238.433
MDL Number MFCD00130144
InChI Key STIIRMZYURVVGK-UHFFFAOYSA-N
PubChem CID 566841
IUPAC Name 3-undecylthiophene
SMILES CCCCCCCCCCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 166°C
Quantity 1 g
Formula Weight 238.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Undecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.