Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Vinylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. V01415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0141-5G 5 g
V01411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0141-5G Supplier TCI America Supplier No. V01415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 3-Vinylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 15016-43-0
Melting Point 145°C
Color White
Molecular Formula C8H9BO2
MDL Number MFCD01074679
Quantity 5 g
Synonym 3-vinylphenylboronic acid, 3-vinylphenyl boronic acid, 3-vinylbenzeneboronic acid, styrene-3-boronic acid, 3-ethenylphenyl boronic acid
InChI Key SYBQEKBVWDPVJM-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C=C)(O)O
IUPAC Name (3-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.968
PubChem CID 4366886
Formula Weight 147.97
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.