Learn More
Caresfield 3-Wall Biohazard Specimen Bag with Tear-Away Zip-Top Closure

Description
- Three-wall construction provides organized specimen containment
- Tear-away design supports convenient access
- Zip-top closure helps keep contents securely contained
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Green |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Latex Free | Yes |
| No. per Pack | 1000/Cs. |
| Quantity | 1000/Cs. |
| Dimensions (L x W) | 10 x 8 in. |
| Length (English) | 10 in. |
| Width (English) | 8 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.