Learn More
SAI Infusion Technologies 3 Way Stopcock - 2 female luers and 1 male luer
SDP
Catalog No. NC0209042
Encompass
Stopcock for directing fluid flow in infusion studies. 3-way stopcock with swivel nut; 200 psi., 2 female luers and one male luer. 10/bx. Product is for laboratory animal research only.
Catalog No. NC0209042 Supplier SAI Infusion Technologies Supplier No. SC3LLHP
Stopcock for directing fluid flow in infusion studies. 3-way stopcock with swivel nut; 200 psi., 2 female luers and one male luer. 10/bx. Product is for laboratory animal research only.