Learn More
New Jersey Fluid Systems LLC 3 WAY BALL VALVE
Catalog No. NC9480494
Encompass
Swagelok 41GX series Ball Valve. 3-way pattern; 1/8 in. OD Tube Fitting ends; 2,500 psi max; 316 Stainless Steel
Catalog No. NC9480494 Supplier New Jersey Fluid Systems LLC Supplier No. SS41GXS2
Swagelok 41GX series Ball Valve. 3-way pattern; 1/8 in. OD Tube Fitting ends; 2,500 psi max; 316 Stainless Steel