Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex 3-Way Direct Lift Solenoid Valve; 1/16 24VDC; 30 psi max
Catalog No. NC1643014
Encompass
Rated for 26" Hg vacuum. Maximum fluid temperature is 158¿F (70¿C). All valves have 15" lead wires. Valves are compact and lightweight ¿ mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
Catalog No. NC1643014 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0154012
Rated for 26" Hg vacuum. Maximum fluid temperature is 158¿F (70¿C). All valves have 15" lead wires. Valves are compact and lightweight ¿ mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.