Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Three-Way, Stopcock with Male Luer Lock, Non-Sterile; 10/PK
Catalog No. NC9593742
Encompass
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
Catalog No. NC9593742 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3060002/DUP
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.