Learn More
Andwin Scientific 3-WAY STOP COCK
SDP
Catalog No. NC0399981
Encompass_Preferred
The 3-way stop cock has 2 Female Luer Locks and 1 Male Luer Lock. This product is ideal for use for infusion and invasive blood pressure monitoring.
Catalog No. NC0399981 Supplier Andwin Scientific Supplier No. 99735
The 3-way stop cock has 2 Female Luer Locks and 1 Male Luer Lock. This product is ideal for use for infusion and invasive blood pressure monitoring.