Learn More
Chemglass Life Sciences 3-Way Stopcock, 9.5mm Connections, for CG-1225-J Vacuum Desiccators
SDP
Catalog No. 501311022
Encompass_Preferred
3-Way Stopcock 9.5mm Connections for CG-1225-J Vacuum Desiccators. Convenient three-way stopcock providing consistent and uniform vacuum draw, vacuum release or shutoff. Vacuum connection has an O.D. of 9.5mm
Catalog No. 50-131-1022 Supplier Chemglass Life Sciences Supplier No. CG1225J23
3-Way Stopcock 9.5mm Connections for CG-1225-J Vacuum Desiccators. Convenient three-way stopcock providing consistent and uniform vacuum draw, vacuum release or shutoff. Vacuum connection has an O.D. of 9.5mm