Learn More
Sigma Aldrich Fine Chemicals Biosciences 3-way stopcock female Luer1EA
Catalog No. 502901500
Encompass
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.Made from nickel plated brass or stainless steelTested to 45 psi water pressureHigh pressure stopcocks are rated to 290 psi
Catalog No. 50-290-1500 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAD60111EA
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.Made from nickel plated brass or stainless steelTested to 45 psi water pressureHigh pressure stopcocks are rated to 290 psi