Learn More
World Precision Instrument 4-Way Stopcock, White, 2 Female Luer Locks, Male Luer with Spin Lock
SDP
Catalog No. 50822022
Encompass
  • Polycarbonate (medical grade) body
  • Polyethylene handle 
  • 130 PSI
  • Package of 10
Catalog No. 50-822-022 Supplier World Precision Instrument Supplier No. 1403510
  • Polycarbonate (medical grade) body
  • Polyethylene handle 
  • 130 PSI
  • Package of 10