Learn More
Ace Glass, Inc. 3-WAY TEFZEL VALVE 3-WAY TEFZEL VALVE
SDP
Supplier:  Ace Glass, Inc. 1273039
Encompass
3-way Tefzel tubing connector valve for applications where stream switching, reagent switching, sample removal, or sample injection is desirable. Valve body is machined of single-piece PTFE. Tefzel valves are rated to 500psi and have one moving part per channel for long life and trouble-free operation. 1.5mm I.D. channels meet at the valve center. Zero dead space within the valve. Valve has polypropylene caps with Fluoron elastomer O-Ring seals, for instant connection of 0.5 to 4mm O.D. rigid or flexible tubing. 1/4¿-28 thread at back of caps. Autoclaveable.
Catalog No. 50-977-787