Learn More
Ibidi USA 3 Well Chamber, removable
SDP
Catalog No. NC1264892
Encompass
The 3 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm×60 mm coverslips are provided by ibidi (Cat.# 10811). The special geometry of the 3 Well Chamber, removable provides a structure inside each well, forming a round inner well. The option of placing a round coverslip (Cat.# 10815) into the well allows the user to fill the inner well exclusively, by minimizing the volume and eliminating the meniscus. Glass slide, silicone chamber, sterile, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC1264892 Supplier Ibidi USA Supplier No. 80381
The 3 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm×60 mm coverslips are provided by ibidi (Cat.# 10811). The special geometry of the 3 Well Chamber, removable provides a structure inside each well, forming a round inner well. The option of placing a round coverslip (Cat.# 10815) into the well allows the user to fill the inner well exclusively, by minimizing the volume and eliminating the meniscus. Glass slide, silicone chamber, sterile, 15 pcs./box