Learn More
Cell Biolabs Inc OxiSelect™ 3-Well Comet Assay Slides, 25 slides
Catalog No. NC0569359
Encompass_Preferred
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Slides provide a fast and sensitive way to measure cellular DNA damage. 25 slides of 3 wells each.
Catalog No. NC0569359 Supplier Cell Biolabs Inc Supplier No. STA353
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Slides provide a fast and sensitive way to measure cellular DNA damage. 25 slides of 3 wells each.