Learn More
Chemglass Life Sciences Plate glass, disc, borosilicate, 3in Diameter X 1/8in Thick
SDP
Supplier:  Chemglass Life Sciences CG190611
Encompass_Preferred
Plate glass, disc, borosilicate, 3in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.
Catalog No. CG190611