Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 3mm ID X 5mm OD
SDP
Catalog No. CGQ031002
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 3mm ID X 5mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031002 Supplier Chemglass Life Sciences Supplier No. CGQ031002
Graded Seal, Quartz to Borosilicate, 3mm ID X 5mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.