Learn More
Chemglass Life Sciences Flask, Shake, 1000mL, Deep Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230
SDP
Catalog No. 501214762
Encompass_Preferred
Flask, Shake, 1000mL, Deep Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230. Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Delong style neck is made to fit stainless steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.
Catalog No. 50-121-4762 Supplier Chemglass Life Sciences Supplier No. CLS204606
Flask, Shake, 1000mL, Deep Baffles, Delong Neck, Approx OD x Height (mm): 130 x 230. Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Delong style neck is made to fit stainless steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.