Learn More
Bellco Glass, Inc. 3 X-DP Shake Flask,125mL 25mm Delong Neck, 12/Case
SDP
Catalog No. 501945528
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mLClosure Sizes: 25mmOD x OAH: 66 x 136mmDeLong Style NeckCase Quantity:2543-00125: 12
Catalog No. 50-194-5528 Supplier Bellco Glass, Inc. Supplier No. 254300125
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mLClosure Sizes: 25mmOD x OAH: 66 x 136mmDeLong Style NeckCase Quantity:2543-00125: 12