Learn More
Bellco Glass, Inc. 3 X-DP Baffle Shake Flask,2000mL 38mm Delong Neck
SDP
Catalog No. 501945533
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2LClosure Sizes: 38mmOD x OAH: 158 x 295mmDeLong Style NeckCase Quantity:2543-02000: 3
Catalog No. 50-194-5533 Supplier Bellco Glass, Inc. Supplier No. 254302000
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2LClosure Sizes: 38mmOD x OAH: 158 x 295mmDeLong Style NeckCase Quantity:2543-02000: 3