Learn More
Bellco Glass, Inc. 3 X-DP Baffle Shake Flask,3000mL 38mm Delong Neck
SDP
Catalog No. 501945534
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 3LClosure Sizes: 38mmOD x OAH: 184 x 340mmDeLong Style NeckCase Quantity:2453-03000: 3
Catalog No. 50-194-5534 Supplier Bellco Glass, Inc. Supplier No. 254303000
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 3LClosure Sizes: 38mmOD x OAH: 184 x 340mmDeLong Style NeckCase Quantity:2453-03000: 3