Learn More
Bellco Glass, Inc. 3 X-DP Baffle Shake Flask,300mL 38mm Delong Neck
SDP
Catalog No. 501945530
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 300mLClosure Sizes: 38mmOD x OAH: 88 x 170mmDeLong Style NeckCase Quantity:2543-00300: 12
Catalog No. 50-194-5530 Supplier Bellco Glass, Inc. Supplier No. 254300300
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 300mLClosure Sizes: 38mmOD x OAH: 88 x 170mmDeLong Style NeckCase Quantity:2543-00300: 12