Learn More
Bellco Glass, Inc. 3 X-DP Baffle Shake Flask,50mL 18mm Delong Neck
SDP
Catalog No. 501945527
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mlClosure Size: 18Dimensions: 50x105mmDelong Style NeckCase Quantity: 12
Catalog No. 50-194-5527 Supplier Bellco Glass, Inc. Supplier No. 254300050
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mlClosure Size: 18Dimensions: 50x105mmDelong Style NeckCase Quantity: 12