Learn More
Polysciences, Inc. Glass Beads, 30 to 50um, Light Green Odorless Beads, CAS No.: 65997-17-3, Store at 25 deg.C, Specific gravity: 2.5, 100g
SDP
Catalog No. nc9154970
Encompass
Glass Beads, 30 to 50um, Light Green Odorless Beads, CAS No.: 65997-17-3, Store at 25 deg.C, Specific gravity: 2.5, 100g
Catalog No. NC9154970 Supplier Polysciences, Inc. Supplier No. 18901100
Glass Beads, 30 to 50um, Light Green Odorless Beads, CAS No.: 65997-17-3, Store at 25 deg.C, Specific gravity: 2.5, 100g