Learn More
Bio Rad Laboratories 30% ACRYL-BIS SOLUTION 500ML
Catalog No. NC9386834
Encompass
Electrophoresis Gel Component; 30% Acrylamide/Bis Solution, 37.5:1; Bio-Rad; 500mL; Ready to use; Made from 99.9% pure acrylamide and bis-acrylamide
Catalog No. NC9386834 Supplier Bio Rad Laboratories Supplier No. 1610158
Electrophoresis Gel Component; 30% Acrylamide/Bis Solution, 37.5:1; Bio-Rad; 500mL; Ready to use; Made from 99.9% pure acrylamide and bis-acrylamide