Learn More
VWR INTERNATIONAL LLC 30% BSA SOLUTION 50ML
Catalog No. NC9432403
Encompass
Concentrated 30% BSA solution (Albumin, Bovine,Fraction V - Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).
Catalog No. NC9432403 Supplier VWR INTERNATIONAL LLC Supplier No. 97063626
Concentrated 30% BSA solution (Albumin, Bovine,Fraction V - Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).