Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biochrom Ultrospec™ 30 Cell Density Meter

Catalog No. BTX80300812 Shop All BTX Harvard Products
Change view
Click to view available options
Display Type:
Color Touch Screen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
BTX80300812 Color Touch Screen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX80300812 Supplier Biochrom Ultrospec™ Supplier No. 80300812
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compact portable meter measures cell density in suspension at 600 nm and supports standard curves using absorbance or McFarland standards.

  • Small, portable, and designed for use, the instrument can measure growth rates of E. coli, other bacteria, and yeast
  • Create and save cell specific standard curves to read against again and again
  • Can also be used with McFarland standards
  • Intuitive touch screen for easy plug and play use
  • Battery operated and portable for use where cells are actually cultured
  • Uses LED light source in combination with an advanced fiber optics
  • Hand-held
  • Accepts 10 x 10 mm cuvettes or 16 mm test tubes. 10 to 12 mm test tubes require adapter set
  • Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with formaldehyde or ethylene oxide to sterilize it
  • Save results onboard or export via USB flash drive
  • Compact, low-maintenance device
  • Not recommended for use with mammalian cell lines

Specifications

Type Cell Density Meter
Light Source LED
Display Type Color Touch Screen
Response Time <2 sec.
Automatic Range Selection Yes

Contains internal rechargeable Li-Ion battery

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.