Learn More
Jensen Global Inc 30 GAUGE NEEDLE X 1IN 50/BAG
Catalog No. NC9279044
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Needle, 1 in., 30G, IT Series, Color Coded Based on the International Standard and are Made with Polypropylene Luer Lock Hub with 304 Stainless Steel Tubing, Bonded with a Clear UV Adhesive for Exceptional Strength
Catalog No. NC9279044 Supplier Jensen Global Inc Supplier No. JG301.0X
This item has a minimum qty of 3 per supplier requirements.
Needle, 1 in., 30G, IT Series, Color Coded Based on the International Standard and are Made with Polypropylene Luer Lock Hub with 304 Stainless Steel Tubing, Bonded with a Clear UV Adhesive for Exceptional Strength