Learn More
SUBREX LLC 30 GAUGE NIAG NC NOZZLES 8/PK
Catalog No. NC1879343
Encompass_Preferred
Blacke Standard 30 Gage, no coating, Nickel Silver, 8/pack. Standard medical taper or luer style hubs. High precision. Reduces dispense pump back pressure, improving performance and extending lifespan of existing equipment. Resists clogging and stringing, improves fluid break-off. Convenient color coded hubs.
Catalog No. NC1879343 Supplier SUBREX LLC Supplier No. A1000300308
Blacke Standard 30 Gage, no coating, Nickel Silver, 8/pack. Standard medical taper or luer style hubs. High precision. Reduces dispense pump back pressure, improving performance and extending lifespan of existing equipment. Resists clogging and stringing, improves fluid break-off. Convenient color coded hubs.