Learn More
GFS Chemicals Inc HYDROGEN PEROXIDE, 30%, VERITAS LOW TRACE METALS | 500 ML | GFS CHEMICALS, INC. Item #: 11782 | Bulk #: 3984
SDP
Catalog No. NC1199178
Encompass_Preferred
HYDROGEN PEROXIDE, 30%, VERITAS LOW TRACE METALS | Chemical Formula : H!2O!2 | CAS # : 7722-84-1 | Specifications : Ag 0.1 ppb , Al 0.1 ppb , As 0.1 ppb , Assay 30.0 - 32.0 %, Au 0.1 ppb , B 0.1 ppb , Ba 0.1 ppb , Be 0.1 ppb , Bi 0.1 ppb , Ca 0.1 ppb , Cd 0.1 ppb , Chloride 0.03 ppm , Co 0.1 ppb , Color (APHA) 10 , Cr 0.1 ppb , Cu 0.1 ppb , Fe 0.1 ppb , Free acid 0.6 ueq/g, Ga 0.1 ppb , Ge 0.1 ppb , K 0.1 ppb , Li 0.1 ppb , Mg 0.1 ppb , Mn 0.1 ppb , Mo 0.1 ppb , Na 0.1 ppb , Nb 0.1 ppb , Ni 0.1 ppb , Pb 0.1 ppb , Phosphate (PO4) (ppm) 0.1 ppm , Sb 0.1 ppb , Sn 0.1 ppb , Sr 0.1 ppb , Sulfate (SO4) (ppm) 0.1 ppm , Ta 0.1 ppb , Ti 0.1 ppb , Tl 0.1 ppb , Total Organic Carbon (ppm) 20 ppm , V 0.1 ppb , Zn 0.1 ppb , Zr 0.1 ppb
Catalog No. NC1199178 Supplier GFS Chemicals Inc Supplier No. 11782
HYDROGEN PEROXIDE, 30%, VERITAS LOW TRACE METALS | Chemical Formula : H!2O!2 | CAS # : 7722-84-1 | Specifications : Ag 0.1 ppb , Al 0.1 ppb , As 0.1 ppb , Assay 30.0 - 32.0 %, Au 0.1 ppb , B 0.1 ppb , Ba 0.1 ppb , Be 0.1 ppb , Bi 0.1 ppb , Ca 0.1 ppb , Cd 0.1 ppb , Chloride 0.03 ppm , Co 0.1 ppb , Color (APHA) 10 , Cr 0.1 ppb , Cu 0.1 ppb , Fe 0.1 ppb , Free acid 0.6 ueq/g, Ga 0.1 ppb , Ge 0.1 ppb , K 0.1 ppb , Li 0.1 ppb , Mg 0.1 ppb , Mn 0.1 ppb , Mo 0.1 ppb , Na 0.1 ppb , Nb 0.1 ppb , Ni 0.1 ppb , Pb 0.1 ppb , Phosphate (PO4) (ppm) 0.1 ppm , Sb 0.1 ppb , Sn 0.1 ppb , Sr 0.1 ppb , Sulfate (SO4) (ppm) 0.1 ppm , Ta 0.1 ppb , Ti 0.1 ppb , Tl 0.1 ppb , Total Organic Carbon (ppm) 20 ppm , V 0.1 ppb , Zn 0.1 ppb , Zr 0.1 ppb