Learn More
Agilent Technologies 30 M, 0.53 MM, 1.00 UM, WITH S
Catalog No. NC1926439
Encompass
J&W DB-WAX GC column, 30 m, 0.53 mm, 1.00 um, with smart key, for 8890 GC system
Catalog No. NC1926439 Supplier Agilent Technologies Supplier No. 1257032KEY
J&W DB-WAX GC column, 30 m, 0.53 mm, 1.00 um, with smart key, for 8890 GC system