Learn More
Apacor 30 mL Clean Transport Vials
Catalog No. 22332033
Change view
Click to view available options
Quantity:
250/Pk.
Catalog No. Quantity
22-332-033 250/Pk.
1 options
Catalog No. 22-332-033 Supplier Apacor Supplier No. 149972
Add to Cart
Add to Cart
For Intestinal Parasite Examination
  • Intestinal parasites are confirmed by the identification of cysts, protozoan trophozoites, larvae and helminth eggs
  • Priorities of these clinical laboratories do not always permit the immediate examination of a fresh sample
  • Prompt collection and transportation of these samples cannot always be guaranteed
  • Freezing, refrigeration and/or incubation of samples cannot ensure the full recovery of parasites at all stages of identification
  • Apacor Transportation Vial will preserve the intestinal parasites in the faecal material until such time it can be examined by a qualified parasitologist
Specifications
Description Sterile Tranport Vial without reagent
Quantity 250/Pk.
Volume (Metric) 30 mL