Learn More
VEOLIA WTS 30-ML POLYSULF- 10 VL/PK
Catalog No. NC0534496
Encompass
One pack containing 10 precleaned 30 ml polysulfone vials. Caps and septa must be ordered separately as HMI 90224-01.
Catalog No. NC0534496 Supplier VEOLIA WTS Supplier No. HMI 9030301
One pack containing 10 precleaned 30 ml polysulfone vials. Caps and septa must be ordered separately as HMI 90224-01.