Learn More
PATHOLOGY INNOVATIONS 30 MM EMBEDDING WELL BAR SET
Catalog No. NC0584211
Encompass_Preferred
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The 30MM Well Bar Set comes with One 30 mm well bar, 4 large chucks, two over chuck freezing blocks and 8 dispensing slides. This set is made for outfitting labs that process a high volume of frozen sections with various sized speciments, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.
Catalog No. NC0584211 Supplier PATHOLOGY INNOVATIONS Supplier No. 113
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The 30MM Well Bar Set comes with One 30 mm well bar, 4 large chucks, two over chuck freezing blocks and 8 dispensing slides. This set is made for outfitting labs that process a high volume of frozen sections with various sized speciments, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.