Learn More
Perkin Elmer US LLC 30 nm Ultra Uniform Gold Nanospheres
Catalog No. 502112684
Encompass_Preferred
Concentration of Element includes: Diameter Description Volume 30 nm 1e7 particles/mL, PEG Carboxyl 0.8 kDa 25 mL
Catalog No. 50-211-2684 Supplier Perkin Elmer US LLC Supplier No. N8151034
Concentration of Element includes: Diameter Description Volume 30 nm 1e7 particles/mL, PEG Carboxyl 0.8 kDa 25 mL