Learn More
Fisherbrand™ 30-Place Cardboard Slide Tray, Black Trim
Catalog No. 14127528 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Cardboard
Catalog No. Material
14-127-528 Cardboard
1 options
Catalog No. 14-127-528
Add to Cart
Add to Cart
Safeguard every slide
  • Designed as a convenient solution for storing and transporting microscope slides.
  • Index on the front cover corresponds with the number grid inside for identification.
  • Recessed slide compartments and inside covers for specimen protection.
  • Slides cannot be accessed when cover is closed reducing contamination.
  • Compartment holder is made from a double layer of densely pressed cardboard.
  • Cover is single layer cardboard.
  • Front protection flaps are attached with durable tape.
  • Available in a variety of different color trims.
Specifications
Type Slide Storage Tray
Slide Capacity 30
Color Black
Dimensions (L x W x H) 8.2 x 12.9 x 0.4 in.
Material Cardboard
Sterility Non-sterile