Learn More
Organomation 30 position MULTIVAP Nitrogen Blowdown Evaporator
SDP
Supplier:  Organomation 11830
Encompass_Preferred
The 30 position MULTIVAP nitrogen blowdown evaporator is a high temperature dry block model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which direct the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 5 samples, which allows individual rows to be shut off when evaporating batches of less than 30 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator. The 30 position MULTIVAP parallel evaporator uses a heated aluminum dry block to reach evaporative temperatures ranging from 30°C to 120°C. Sample blocks are customized for the user’s sample size for maximum and even heat transfer. Inserts can be customized for sample vial outside diameters of 10-30 mm.
Catalog No. NC2301751
Add to Cart
Request a Quote