Learn More
Valutek™ 30 Pound Tabloid-Size Cleanroom Paper
Catalog No. 191502134
Change view
Click to view available options
Quantity:
250 Sheets/Ream, 5 Reams/Cs., 1250 Sheets/Cs.
Catalog No. Quantity
19-150-2134 250 Sheets/Ream, 5 Reams/Cs., 1250 Sheets/Cs.
1 options
Catalog No. 19-150-2134 Supplier Valutek™ Supplier No. VTCRPS30B1117
Add to Cart
Add to Cart
Specially formulated with cellulose fiber surrounded and bonded by a polymer impregnated substrate. The special formulation minimizes particulate and ink smearing. This also provides the paper with high tear and tensile strength but with ultra-low levels of particle and extractable counts.
  • 250 sheets/ream, 5 reams/case, 1250 sheets/case
  • Ultra-low levels of particle and extractable counts
  • This product is commonly used for note-taking, laser printing, and photocopies. It is also used as a form and log sheet in a cleanroom Class 1 -10 (ISO 3-4) critical environment
  • It is compatible with most inks without smearing while ensuring low particle and fiber generation and low ionic contamination
  • All Valutek™ cleanroom papers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
Specifications
Material Cellulose
Color Blue
Cover Material Polymer impregnated substrate
Dimensions (L x W) 11 x 17 in.
Quantity 250 Sheets/Ream, 5 Reams/Cs., 1250 Sheets/Cs.