Learn More
Electron Microscopy Sciences 30-Slide Staining Dish Complete Set, 3¾" x 3⅞" x 3-3/8" (H)
SDP
Catalog No. NC1603646
Encompass_Preferred
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Includes one staining dish with lid and one handle rack. Accomodates 30 slides.
I.D.: 3¾" x 3⅞" x 3-3/8" (H)
Dimensions: 4" x 4⅛" x 4" (H)
Catalog No. NC1603646 Supplier Electron Microscopy Sciences Supplier No. 7143025
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Includes one staining dish with lid and one handle rack. Accomodates 30 slides.
I.D.: 3¾" x 3⅞" x 3-3/8" (H)
Dimensions: 4" x 4⅛" x 4" (H)