Learn More
Electron Microscopy Sciences 30-Slide Staining Dish Set
SDP
Catalog No. 5028704
Encompass_Preferred
Set contains 1 staining dish, 1 lid, and 1 30-slide stainless steel rack. This staining dish is made from soda lime and accommodates 16, 20 and 30-slide racks. Slides range from 3" x 1" (76 x 25 mm), 3" x 1 1/2" (76 x 38 mm), and 3 x 2" (76 x 51 mm), and 3" x 2" (76 x 51 mm). The approximate inside dimensions are: 135H x 102W x 69D mm. The out side dimensions are 146H x 113W x 83D mm. Contains 1 dish, 1 rack, 1 cover.
Catalog No. 50-287-04 Supplier Electron Microscopy Sciences Supplier No. 7031230
Set contains 1 staining dish, 1 lid, and 1 30-slide stainless steel rack. This staining dish is made from soda lime and accommodates 16, 20 and 30-slide racks. Slides range from 3" x 1" (76 x 25 mm), 3" x 1 1/2" (76 x 38 mm), and 3 x 2" (76 x 51 mm), and 3" x 2" (76 x 51 mm). The approximate inside dimensions are: 135H x 102W x 69D mm. The out side dimensions are 146H x 113W x 83D mm. Contains 1 dish, 1 rack, 1 cover.