Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ 300 g Electronic Balance UltraClass Weight

Catalog No. 02218085 Shop All Troemner Products
Change view
Click to view available options
Certifications/Compliance:
No Certificate
Traceable Certificate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
02-218-085 No Certificate
02-218-086 Traceable Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-218-085 Supplier Troemner™ Supplier No. 30390283
Add to Cart
Add to Cart

Combine high precision with the advantage of two-piece construction. UltraClass weights, exclusive to Troemner™ are the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • Supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
TRUSTED_SUSTAINABILITY

Specifications

Product Type Two-Piece Analytical Weight
Certifications/Compliance No Certificate
Classification UltraClass
Material Stainless Steel
Case Material Polycarbonate
Density 7.95 g/cm3 at 20°C
Mass 300 g
Tolerance 0.38 mg
Length (Metric) 8.89 cm
Height (Metric) Case 11.43 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Cylindrical Weight, Polycarbonate Case, Glove
Quantity Each
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.