Learn More
Analytical Sales & Services 300µL Glass Inserts with Bottom Spring - for 15 x 45 Vials
Catalog No. 5000806427
Encompass
Advantage™ 300µL Volume, Glass Inserts with Bottom Spring - for 15 x 45 Vials
Catalog No. 50-008-06427 Supplier Analytical Sales & Services Supplier No. 30501CASE
Advantage™ 300µL Volume, Glass Inserts with Bottom Spring - for 15 x 45 Vials