Learn More
Sentry Air Systems Model 300 Portable Fume Extractor
SDP
Catalog No. NC2382247
Encompass
The Portable Fume Extractor provides a vital engineering control for medical and laboratory facilities. The 68 in. (172.72 cm) long capture arm can easily be moved to capture chemical fumes, vapors, and particulate at the source. This model includes a portability handle, heavy-duty casters, a variable speed control, and an easy to change filter system. Both activated carbon and particulate filters can be housed in the filtration chamber to capture both fumes and particulate. Approximate Airflow: up to 350 CFM. Filters Available: HEPA Filter, ASHRAE Filter, ULPA Filter, Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].
Catalog No. NC2382247 Supplier Sentry Air Systems Supplier No. SS300PFS
The Portable Fume Extractor provides a vital engineering control for medical and laboratory facilities. The 68 in. (172.72 cm) long capture arm can easily be moved to capture chemical fumes, vapors, and particulate at the source. This model includes a portability handle, heavy-duty casters, a variable speed control, and an easy to change filter system. Both activated carbon and particulate filters can be housed in the filtration chamber to capture both fumes and particulate. Approximate Airflow: up to 350 CFM. Filters Available: HEPA Filter, ASHRAE Filter, ULPA Filter, Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].