Learn More
Chemglass Life Sciences Chemglass 300 μL Automation Clear Pipette Tips for Hamilton Systems - Sterile, Boxed (96/rack, 10 racks/pack)
SDP
Catalog No. 502733258
Encompass_Preferred
Designed for high-throughput automated pipetting, these 300 μL clear automation tips are supplied sterile with barcodes and manufactured from medical-grade polypropylene for contamination-controlled sample handling.
  • 300 μL capacity.
  • Medical-grade polypropylene construction.
  • Sterile; electron beam sterilization.
  • Box-packed; 96 per rack, 10 racks per pack, 50 racks per case.
  • Barcode on each rack for inventory tracking.
  • DNase/RNase free and produced in a 100,049 grade clean room.
  • Intended for high-throughput workflows in genomics, proteomics, immunoassays, and biopharmaceutical research.
Catalog No. 50-273-3258 Supplier Chemglass Life Sciences Supplier No. CLS3499A063
Designed for high-throughput automated pipetting, these 300 μL clear automation tips are supplied sterile with barcodes and manufactured from medical-grade polypropylene for contamination-controlled sample handling.
  • 300 μL capacity.
  • Medical-grade polypropylene construction.
  • Sterile; electron beam sterilization.
  • Box-packed; 96 per rack, 10 racks per pack, 50 racks per case.
  • Barcode on each rack for inventory tracking.
  • DNase/RNase free and produced in a 100,049 grade clean room.
  • Intended for high-throughput workflows in genomics, proteomics, immunoassays, and biopharmaceutical research.