Learn More
Chemglass Life Sciences 300 μl Automation Conductive Filter Tips for Hamilton systems - Sterile, conductive, filtered, barcode, 96 tips x 5 blisters/pack
SDP
Catalog No. 502733276
Encompass_Preferred
300 μl automation conductive filter tips are sterile, barcode-equipped pipette tips engineered for high-throughput liquid handling on Hamilton systems. They combine aerosol-resistant filtration and a conductive design to support reliable liquid level detection, reduce contamination risk, and simplify automated workflows in genomics, proteomics, and other laboratory applications.
  • Tip volume: 300 μl.
  • Compatible with Hamilton systems.
  • Electron beam sterilized and DNase/RNase free.
  • Aerosol-resistant filter to reduce contamination.
  • Conductive tip design for liquid level detection.
  • Packaging: 96 tips x 5 blisters per pack; 5,760 tips per case.
  • Made from medical-grade polypropylene.
  • Manufactured in a 100,049 grade clean room.
Catalog No. 50-273-3276 Supplier Chemglass Life Sciences Supplier No. CLS3499A023
300 μl automation conductive filter tips are sterile, barcode-equipped pipette tips engineered for high-throughput liquid handling on Hamilton systems. They combine aerosol-resistant filtration and a conductive design to support reliable liquid level detection, reduce contamination risk, and simplify automated workflows in genomics, proteomics, and other laboratory applications.
  • Tip volume: 300 μl.
  • Compatible with Hamilton systems.
  • Electron beam sterilized and DNase/RNase free.
  • Aerosol-resistant filter to reduce contamination.
  • Conductive tip design for liquid level detection.
  • Packaging: 96 tips x 5 blisters per pack; 5,760 tips per case.
  • Made from medical-grade polypropylene.
  • Manufactured in a 100,049 grade clean room.