Learn More
Hoefer, Inc Hoefer™ PS300B 300 Volt Power Supply
SDP
Catalog No. NC2399102
Encompass_Preferred
The versatile PS300B is designed for most electrophoresis techniquesFeatures:Constant voltage or constant current mode with automatic crossover10 to 300 V, 4 to 500 mA, 90 WFour sets of 4 mm output jacksIdeal choice for most electrophoresis techniquesContinuous runs or 999 minute timerAutomatic recovery after power failureOpen circuit detectionShort circuit protectionModes of Operation:Constant Voltage Mode—the power supply will attempt to maintain the user selected voltage by automatically adjusting the current flowConstant Current Mode—the power supply will attempt to maintain the user selected current flow by automatically adjusting the voltage suppliedAutomatic Crossover—when the constant parameter (voltage or current) cannot be maintained due to power supply operating limits the power supply automatically switches to maintaining the other parameter
Catalog No. NC2399102 Supplier Hoefer, Inc Supplier No. PS300B
The versatile PS300B is designed for most electrophoresis techniquesFeatures:Constant voltage or constant current mode with automatic crossover10 to 300 V, 4 to 500 mA, 90 WFour sets of 4 mm output jacksIdeal choice for most electrophoresis techniquesContinuous runs or 999 minute timerAutomatic recovery after power failureOpen circuit detectionShort circuit protectionModes of Operation:Constant Voltage Mode—the power supply will attempt to maintain the user selected voltage by automatically adjusting the current flowConstant Current Mode—the power supply will attempt to maintain the user selected current flow by automatically adjusting the voltage suppliedAutomatic Crossover—when the constant parameter (voltage or current) cannot be maintained due to power supply operating limits the power supply automatically switches to maintaining the other parameter