Learn More
Edmund Optics 300 x 300mm, λ/4 Retarder Film (WP140HE)
Catalog No. NC2750428
Encompass
Polymer Retarder Film is an extremely durable, birefringent polymer sheet that has been designed to modify the polarization state of transmitted light. The highly transparent, achromatic λ/4 (WP140HE) Polymer Retarder Film consists of a single layer, modified polycarbonate sheet that acts an achromatic retarder, and is optically and mechanically sound up to 90°C or 80°C respectively. Ideal for use in a variety of coherent and polarized light applications, including circular polarizers, LCDs, and fiber optics.
Catalog No. NC2750428 Supplier Edmund Optics Supplier No. 14725
Polymer Retarder Film is an extremely durable, birefringent polymer sheet that has been designed to modify the polarization state of transmitted light. The highly transparent, achromatic λ/4 (WP140HE) Polymer Retarder Film consists of a single layer, modified polycarbonate sheet that acts an achromatic retarder, and is optically and mechanically sound up to 90°C or 80°C respectively. Ideal for use in a variety of coherent and polarized light applications, including circular polarizers, LCDs, and fiber optics.