Learn More
CHEMSCIENCE Serialized and Certified, Volumetric Flask, Low Actinic Amber, 3000mL, Wide Mouth, PE Stopper#32, Class A
Catalog No. NC2307539
Encompass_Preferred
Serialized and Certified, Volumetric Flask, Low Actinic Amber, 3000mL, Wide Mouth, PE Stopper#32, Class A
Catalog No. NC2307539 Supplier CHEMSCIENCE Supplier No. 130.502.11
Serialized and Certified, Volumetric Flask, Low Actinic Amber, 3000mL, Wide Mouth, PE Stopper#32, Class A