Learn More
Cole-Parmer Cole-Parmer Low-Form Polypropylene Beakers with Handle and Pour Spout, 3000 mL, 2/Pk
Catalog No. NC0719132 Shop All Cole-Parmer Products
Encompass_Preferred
Low-form design adds stability to these beakers. Made of polypropylene (PP), beakers are ideal for many industrial applications. Graduated in milliliters and liters.These beakers can withstand temperatures to 275F (135C).
Catalog No. NC0719132 Supplier Cole-Parmer Supplier No. 6100052
Add to Cart
Add to Cart
Low-form design adds stability to these beakers. Made of polypropylene (PP), beakers are ideal for many industrial applications. Graduated in milliliters and liters.These beakers can withstand temperatures to 275F (135C).