Learn More
Chemglass Life Sciences 3000mL Glassblowers Erlenmeyer Blank
SDP
Catalog No. CG62011
Encompass_Preferred
3000mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG62011 Supplier Chemglass Life Sciences Supplier No. CG62011
3000mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.