Learn More
Ace Glass, Inc. Separatory funnel, 3000mL, 45/50, 6mm bore glass stopcock
SDP
Catalog No. 50974674
Encompass
3000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 45/50 standard taper top joint, and a 6mm bore glass stopcock plug.
Catalog No. 50-974-674 Supplier Ace Glass, Inc. Supplier No. 722422
3000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 45/50 standard taper top joint, and a 6mm bore glass stopcock plug.